Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAPZA1 Antibody (OTI2G4), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270336MFV500
Description
CAPZA1 Monoclonal antibody specifically detects CAPZA1 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
CAPZA1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI2G4 | |
Flow Cytometry | |
Cap Z, CAPPA1, capping protein (actin filament) muscle Z-line, alpha 1, CAPZ, CapZ alpha-1, CAZ1, F-actin-capping protein subunit alpha-1 | |
Full length human recombinant protein of human CAPZA1(NP_006126) produced in HEK293T cell. | |
0.1 mL | |
Extracellular Matrix | |
829 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction