Learn More
Invitrogen™ CAPZA1 Monoclonal Antibody (OTI2G4)

Description
CAPZA1 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | CAPZA1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2G4 |
| Concentration | 1.00 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CAPZA1 |
| Gene Accession No. | P52907 |
| Gene Alias | Cap Z; Cappa1; capping actin protein of muscle Z-line alpha subunit 1; capping actin protein of muscle Z-line subunit alpha 1; capping protein (actin filament) muscle Z-line, alpha 1; capping protein alpha 1; CAPZ; CapZ alpha-1; capZ alpha-1 {ECO:0000250; CAPZA1; capza1 {ECO:0000250; CAZ1; F-actin capping protein alpha-1 subunit; F-actin-capping protein subunit alpha-1; F-actin-capping protein subunit alpha-1 {ECO:0000250; UniProtKB:P52907}; unnamed protein product |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.