Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPZA1 Mouse anti-Human, Mouse, Rat, Clone: 10H1F9, Proteintech
SDP

Catalog No. 501736503 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6503 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6503 Supplier Proteintech Group Inc Supplier No. 660661IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CAPZA1 is a member of the F-actin capping protein alpha subunit family. This gene encodes the alpha subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.

Specifications

Antigen CAPZA1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 10H1F9
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CAPZA1
Gene Accession No. B2GUZ5, P47753, P52907
Gene Alias CAPPA1, CAPZ, CapZ alpha 1, CAPZA1, CAZ1
Gene Symbols CAPZA1
Host Species Mouse
Immunogen CAPZA1 Fusion Protein Ag18841
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12340, 691149, 829
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.