Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAPZA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531026
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531026 Supplier Invitrogen™ Supplier No. PA531026
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji, NIH-3T3. Predicted reactivity: Mouse (95%), Rat (96%), Xenopus laevis (81%), Chicken (89%), Rhesus Monkey (99%), Chimpanzee (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CAPZA1 is a member of the F-actin capping protein alpha subunit family. This gene encodes the alpha subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAPZA1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene CAPZA1
Gene Accession No. P47753, P52907
Gene Alias Cap Z; Cappa1; capping actin protein of muscle Z-line alpha subunit 1; capping actin protein of muscle Z-line subunit alpha 1; capping protein (actin filament) muscle Z-line, alpha 1; capping protein alpha 1; CAPZ; CapZ alpha-1; capZ alpha-1 {ECO:0000250; CAPZA1; capza1 {ECO:0000250; CAZ1; F-actin capping protein alpha-1 subunit; F-actin-capping protein subunit alpha-1; F-actin-capping protein subunit alpha-1 {ECO:0000250; UniProtKB:P52907}; unnamed protein product
Gene Symbols CAPZA1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CAPZA1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12340, 829
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.