Learn More
Invitrogen™ CAPZA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596255
Description
Immunogen sequence: MADFDDRVSD EEKVRIAAKF ITHAPPGEFN EVFNDVRLLL NNDNLLREGA AHAFAQYNMD QFTPVKIEGY EDQVLITEHG DLGNSRFLDP RNKISFKFDH LRKEASDPQP EEADGGLKSW RESCDSALRA YVKDHYSNGF CTVYAKTIDG QQTIIACIES HQFQPKNFWN GRWRSEWKFT ITPPTAQVVG VLKIQVHYYE DGNVQLVSHK DVQDSLTVSN EAQTAKEFIK IIENAENEYQ TAISENYQTM SDTTFKALRR QLPVTRTKID WNKILSYKIG KEMQNA; Positive Samples: HeLa, Jurkat, MCF7, HL-60, Mouse brain, Mouse kidney, Mouse lung, Rat lung; Cellular Location: Cytoplasm, cytoskeleton.
CAPZA1 is a member of the F-actin capping protein alpha subunit family. This gene encodes the alpha subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.
Specifications
CAPZA1 | |
Polyclonal | |
Unconjugated | |
CAPZA1 | |
Cap Z; Cappa1; capping actin protein of muscle Z-line alpha subunit 1; capping actin protein of muscle Z-line subunit alpha 1; capping protein (actin filament) muscle Z-line, alpha 1; capping protein alpha 1; CAPZ; CapZ alpha-1; capZ alpha-1 {ECO:0000250; CAPZA1; capza1 {ECO:0000250; CAZ1; F-actin capping protein alpha-1 subunit; F-actin-capping protein subunit alpha-1; F-actin-capping protein subunit alpha-1 {ECO:0000250; UniProtKB:P52907}; unnamed protein product | |
Rabbit | |
Affinity chromatography | |
RUO | |
12340, 691149, 829 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.53 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B2GUZ5, P47753, P52907 | |
CAPZA1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-286 of human CAPZA1 (NP_0061261). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.