Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAPZA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529982 Supplier Invitrogen™ Supplier No. PA529982
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2, HCT116, NIH-3T3. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (94%), Dog (100%), Cat (100%), Pig (100%), Rabbit (97%), Chicken (97%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the F-actin capping protein alpha subunit family. It is the alpha subunit of the barbed-end actin binding protein Cap Z. By capping the barbed end of actin filaments, Cap Z regulates the growth of the actin filaments at the barbed end.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAPZA2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Capza2
Gene Accession No. P47754, P47755
Gene Alias 1110053K06Rik; AW208808; Cappa2; capping actin protein of muscle Z-line alpha subunit 2; capping protein (actin filament) muscle Z-line, alpha 2; capping protein alpha 2; CAPZ; capZ alpha-2; CAPZA2; CAZA2; F-actin capping protein alpha-2 subunit; F-actin-capping protein subunit alpha-2
Gene Symbols Capza2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CAPZA2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12343, 830
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.