Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAPZA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117832
This item is not returnable.
View return policy
Description
CAPZA2 Polyclonal Antibody for Western Blot, IP
The protein encoded by this gene is a member of the F-actin capping protein alpha subunit family. It is the alpha subunit of the barbed-end actin binding protein Cap Z. By capping the barbed end of actin filaments, Cap Z regulates the growth of the actin filaments at the barbed end.
Specifications
| CAPZA2 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| P47755 | |
| Capza2 | |
| E. coli-derived Human CAPZA2 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Capza2 | |
| 1110053K06Rik; AW208808; Cappa2; capping actin protein of muscle Z-line alpha subunit 2; capping protein (actin filament) muscle Z-line, alpha 2; capping protein alpha 2; CAPZ; capZ alpha-2; CAPZA2; CAZA2; F-actin capping protein alpha-2 subunit; F-actin-capping protein subunit alpha-2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 830 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction