Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAPZA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120748
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120748 Supplier Invitrogen™ Supplier No. PA5120748
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, THP-1, 22Rv1, Mouse brain, Mouse heart. Immunogen sequence: MADLEEQLSD EEKVRIAAKF IIHAPPGEFN EVFNDVRLLL NNDNLLREGA AHAFAQYNLD QFTPVKIEGY EDQVLITEHG DLGNGKFLDP KNRICFKFDH LRKEATDPRP CEVENAVESW RTSVETALRA YVKEHYPNGV CTVYGKKIDG QQTIIACIES HQFQAKNFWN GRWRSEWKFT ITPSTTQVVG ILKIQVHYYE DGNVQLVSHK DIQDSLTVSN EVQTAKEFIK IVEAAENEYQ TAISENYQTM SDTTFKALRR QLPVTRTKID WNKILSYKIG KEMQNA.

The protein encoded by this gene is a member of the F-actin capping protein alpha subunit family. It is the alpha subunit of the barbed-end actin binding protein Cap Z. By capping the barbed end of actin filaments, Cap Z regulates the growth of the actin filaments at the barbed end.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAPZA2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Capza2
Gene Accession No. P47754, P47755
Gene Alias 1110053K06Rik; AW208808; Cappa2; capping actin protein of muscle Z-line alpha subunit 2; capping protein (actin filament) muscle Z-line, alpha 2; capping protein alpha 2; CAPZ; capZ alpha-2; CAPZA2; CAZA2; F-actin capping protein alpha-2 subunit; F-actin-capping protein subunit alpha-2
Gene Symbols Capza2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-286 of human CAPZA2 (NP_006127.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12343, 830
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.