Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAPZA3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CAPZA3 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CAPZA3 Polyclonal specifically detects CAPZA3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CAPZA3 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CAPAA3, CAPPA3F-actin capping protein alpha-3 subunit, capping protein (actin filament) muscle Z-line, alpha 3, CapZ alpha-3, CP-alpha-3, F-actin-capping protein subunit alpha-3, Germ cell-specific protein 3, GSG3 | |
CAPZA3 | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
Polyclonal | |
Rabbit | |
Human | |
Q96KX2 | |
93661 | |
This antibody was developed against a recombinant protein corresponding to amino acids: KDKERVIRRLLLQAPPGEFVNAFDDLCLLIRDEKLMHHQGECAGHQHCQKYSVPLCIDGNPVLLSHHNVMGDYRFFDHQSKLSFKYDLLQN | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title