Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAPZA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558494
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558494 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558494 Supplier Invitrogen™ Supplier No. PA558494
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KDKERVIRRL LLQAPPGEFV NAFDDLCLLI RDEKLMHHQG ECAGHQHCQK YSVPLCIDGN PVLLSHHNVM GDYRFFDHQS KLSFKYDLLQ N Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

This gene encodes an actin capping protein, one of the F-actin capping protein alpha subunit family. The encoded protein is predominantly localized to the neck region of ejaculated sperm, other immunohistochemical signals were found in the tail and postacrosomal regions. The encoded protein may also form heterodimers of alpha and beta subunits. This protein may be important in determining sperm architecture and male fertility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAPZA3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAPZA3
Gene Accession No. Q96KX2
Gene Alias 510-4; CAPAA3; Cappa3; capping actin protein of muscle Z-line alpha subunit 3; capping actin protein of muscle Z-line subunit alpha 3; capping protein (actin filament) muscle Z-line, alpha 3; capping protein alpha 3; capZ alpha-3; Capza3; cp alpha3; CP-alpha-3; epididymis secretory protein Li 86; F-actin capping protein alpha-3 subunit; F-actin-capping protein subunit alpha-3; Germ cell-specific protein 3; GSG3; HEL-S-86; repro32; testis tissue sperm-binding protein Li 73n; Tex8
Gene Symbols CAPZA3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CAPZA3. Recombinant protein control fragment (Product #RP-102845).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93661
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.