Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPZA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570405 Supplier Thermo Scientific Supplier No. PA570405
Add to Cart
Add to Cart

CAPZA3 Polyclonal Antibody

CAPZA3 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CAPZA3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CAPZA3
Gene Accession No. Q96KX2
Gene Alias 510-4; CAPAA3; Cappa3; capping actin protein of muscle Z-line alpha subunit 3; capping actin protein of muscle Z-line subunit alpha 3; capping protein (actin filament) muscle Z-line, alpha 3; capping protein alpha 3; capZ alpha-3; Capza3; cp alpha3; CP-alpha-3; epididymis secretory protein Li 86; F-actin capping protein alpha-3 subunit; F-actin-capping protein subunit alpha-3; Germ cell-specific protein 3; GSG3; HEL-S-86; repro32; testis tissue sperm-binding protein Li 73n; Tex8
Gene Symbols CAPZA3
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human CAPZA3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93661
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.