Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAPZB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565996
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565996 Supplier Invitrogen™ Supplier No. PA565996
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GCWDSIHVVEV QEKSSGRTAH YKLTSTVMLW LQTNKSGSGT MNLGGSLTRQ MEKDETVSDC SP Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

CAPZB is a member of the F-actin capping protein family. This gene encodes the beta subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAPZB
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAPZB
Gene Accession No. P47756
Gene Alias 1700120C01Rik; AI325129; Cap Z; CAPB; CAPPB; Cappb1; capping actin protein of muscle Z-line beta subunit; capping actin protein of muscle Z-line subunit beta; capping protein (actin filament) muscle Z-line, beta; capping protein beta 1; CAPZ; capZ beta; Capzb; CP beta1; CPB1; CPB2; CPbeat2; CPbeta1; CPbeta2; F-actin capping protein beta subunit; F-actin capping protein subunit beta 1; F-actin-capping protein subunit beta; MGC104401; MGC129749; MGC129750; RP4-657E11.7
Gene Symbols CAPZB
Host Species Rabbit
Immunogen Recombinant Human CAPZB. Recombinant protein control fragment (Product #RP-105031).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 832
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.