Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAPZB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596935
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596935 Supplier Invitrogen™ Supplier No. PA596935
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSDQQLDCAL DLMRRLPPQQ IEKNLSDLID LVPSLCEDLL SSVDQPLKIA RDKVVGKDYL LCDYNRDGDS YRSPWSNKYD PPLEDGAMPS ARLRKLEVEA NNAFDQYRDL YFEGGVSSVY LWDLDHGFAG VILIKKAGDG SKKIKGCWDS IHVVEVQEKS SGRTAHYKLT STVMLWLQTN KSGSGTMNLG GSLTRQMEKD ETVSDCSPHI ANIGRLVEDM ENKIRSTLNE IYFGKTKDIV NGLRSVQTFA DKSKQEALKN DLVEALKRKQ QC; Positive Samples: A-431, THP-1, HeLa, Mouse brain, Mouse testis, Mouse spleen, Rat small intestine; Cellular Location: Cytoplasm, cytoskeleton, myofibril, sarcomere.

CAPZB is a member of the F-actin capping protein family. This gene encodes the beta subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAPZB
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CAPZB
Gene Accession No. P47756, P47757, Q5XI32
Gene Alias 1700120C01Rik; AI325129; Cap Z; CAPB; CAPPB; Cappb1; capping actin protein of muscle Z-line beta subunit; capping actin protein of muscle Z-line subunit beta; capping protein (actin filament) muscle Z-line, beta; capping protein beta 1; CAPZ; capZ beta; Capzb; CP beta1; CPB1; CPB2; CPbeat2; CPbeta1; CPbeta2; F-actin capping protein beta subunit; F-actin capping protein subunit beta 1; F-actin-capping protein subunit beta; MGC104401; MGC129749; MGC129750; RP4-657E11.7
Gene Symbols CAPZB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-272 of human CAPZB (NP_0049211).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12345, 298584, 832
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.