Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPZB Polyclonal Antibody, Huabio

Catalog No. 502544537
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4537 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4537 Supplier HUABIO RESEARCH INC Supplier No. HA500486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CAPZB Polyclonal antibody specifically detects CAPZB in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen CAPZB
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene CAPZB
Gene Accession No. P47756, P47757
Gene Alias 1700120C01Rik; AI325129; Cap Z; CAPB; CAPPB; Cappb1; capping actin protein of muscle Z-line beta subunit; capping actin protein of muscle Z-line subunit beta; capping protein (actin filament) muscle Z-line, beta; capping protein beta 1; CAPZ; capZ beta; Capzb; CP beta1; CPB1; CPB2; CPbeat2; CPbeta1; CPbeta2; F-actin capping protein beta subunit; F-actin capping protein subunit beta 1; F-actin-capping protein subunit beta; MGC104401; MGC129749; MGC129750; RP4-657E11.7
Host Species Rabbit
Immunogen Synthetic peptide within human CAPZB aa 227-277/277.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12345, 832
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.