Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAR/NR1I3 Antibody (OTI1C6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271525H
Description
CAR/NR1I3 Monoclonal specifically detects CAR/NR1I3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CAR/NR1I3 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
NR1I3 | |
Recombinant protein fragment corresponding to amino acids 1-236 of human CAR/NR1I3 (NP_001070950) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1C6 | |
Western Blot, Immunohistochemistry | |
CARCAR1, Constitutive activator of retinoid response, constitutive active receptor, Constitutive active response, Constitutive androstane receptor, MB67, MGC150433, MGC97144, MGC97209, nuclear receptor subfamily 1 group I member 3, nuclear receptor subfamily 1, group I, member 3, orphan nuclear hormone receptor, Orphan nuclear receptor MB67 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
9970.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction