Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAR Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50 - $596.50
Specifications
Antigen | CAR |
---|---|
Concentration | 0.2 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5110995
|
Invitrogen™
PA5110995 |
100 μL |
Each for $596.50
|
|
|||||
PIPA5110996
|
Invitrogen™
PA5110996 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: EVHHDIREDV PPPKSRTSTA RSYIGSNHSS LGSMSPSNME GYSKTQYNQV PSEDFERTPQ SPTLPPAKVA APNLS.
The protein encoded by this gene is a type I membrane receptor for group B coxsackieviruses and subgroup C adenoviruses. Pseudogenes of this gene are found on chromosomes 15, 18, and 21.Specifications
CAR | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
CXADR | |
2610206D03Rik; 46 kD coxsackievirus and adenovirus receptor (CAR) protein; AU016810; AW553441; CAR; Car1; CAR4/6; coxsackie virus and adenovirus receptor; coxsackievirus and adenovirus receptor; Coxsackievirus and adenovirus receptor homolog; coxsackievirus B-adenovirus receptor; CVB3-binding protein; Cxadr; HCAR; HCVADR; MCAR; MCVADR; rCAR | |
CXADR | |
Primary | |
Antigen affinity chromatography |
0.2 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P78310 | |
1525 | |
Recombinant Protein corresonding to Human CAR. Recombinant protein control fragment (Product #RP-95888). | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title