Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595942 Supplier Invitrogen™ Supplier No. PA595942
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSITTPEEMI EKAKGETAYL PCKFTLSPED QGPLDIEWLI SPADNQKVDQ VIILYSGDKI YDDYYPDLKG RVHFTSNDLK SGDASINVTN LQLSDIGTYQ CKVKKAPGVA NKKIHLVVLV KPSGARCYVD GSEEIGSDFK IKCEPKEGSL PLQYEWQKLS DSQKMPTSWL AEMTSSVISV KNASSEYSGT YSCTVRNRVG SDQCLLRLNV VPPSNKAG; Positive Samples: HeLa, OVCAR3, SW480, HepG2, PC-3, Mouse liver, Mouse heart; Cellular Location: Basolateral cell membrane, Cell junction, Cell membrane, Single-pass membrane protein, Single-pass type I membrane protein, Single-pass type I membrane protein, adherens junction, tight junction.

The protein encoded by this gene is a type I membrane receptor for group B coxsackieviruses and subgroup C adenoviruses. Pseudogenes of this gene are found on chromosomes 15, 18, and 21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAR
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CXADR
Gene Accession No. P78310, P97792, Q9R066
Gene Alias 2610206D03Rik; 46 kD coxsackievirus and adenovirus receptor (CAR) protein; AU016810; AW553441; CAR; Car1; CAR4/6; coxsackie virus and adenovirus receptor; coxsackievirus and adenovirus receptor; Coxsackievirus and adenovirus receptor homolog; coxsackievirus B-adenovirus receptor; CVB3-binding protein; Cxadr; HCAR; HCVADR; MCAR; MCVADR; rCAR
Gene Symbols CXADR
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-237 of human CAR/CXADR (NP_0013291).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13052, 1525, 89843
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.