Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110995
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110995 100 μL
PIPA5110996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5110995 Supplier Invitrogen™ Supplier No. PA5110995
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVHHDIREDV PPPKSRTSTA RSYIGSNHSS LGSMSPSNME GYSKTQYNQV PSEDFERTPQ SPTLPPAKVA APNLS Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 88%.

The protein encoded by this gene is a type I membrane receptor for group B coxsackieviruses and subgroup C adenoviruses. Pseudogenes of this gene are found on chromosomes 15, 18, and 21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAR
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CXADR
Gene Accession No. P78310
Gene Alias 2610206D03Rik; 46 kD coxsackievirus and adenovirus receptor (CAR) protein; AU016810; AW553441; CAR; Car1; CAR4/6; coxsackie virus and adenovirus receptor; coxsackievirus and adenovirus receptor; Coxsackievirus and adenovirus receptor homolog; coxsackievirus B-adenovirus receptor; CVB3-binding protein; Cxadr; HCAR; HCVADR; MCAR; MCVADR; rCAR
Gene Symbols CXADR
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CAR. Recombinant protein control fragment (Product #RP-95888).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1525
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.