Learn More
Invitrogen™ CAR Recombinant Rabbit Monoclonal Antibody (068)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA530473
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human CXADR.
The protein encoded by this gene is a type I membrane receptor for group B coxsackieviruses and subgroup C adenoviruses. Pseudogenes of this gene are found on chromosomes 15, 18, and 21.
Specifications
CAR | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7 | |
P78310 | |
CXADR | |
Recombinant Human CXADR protein (Met1-Gly237). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
068 | |
Unconjugated | |
CXADR | |
2610206D03Rik; 46 kD coxsackievirus and adenovirus receptor (CAR) protein; AU016810; AW553441; CAR; Car1; CAR4/6; coxsackie virus and adenovirus receptor; coxsackievirus and adenovirus receptor; Coxsackievirus and adenovirus receptor homolog; coxsackievirus B-adenovirus receptor; CVB3-binding protein; Cxadr; HCAR; HCVADR; MCAR; MCVADR; rCAR | |
Rabbit | |
Protein A | |
RUO | |
1525 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.