Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
carabin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215695
Description
carabin Polyclonal specifically detects carabin in Human samples. It is validated for Western Blot.Specifications
carabin | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
carabin, FLJ00332, MGC46488, TBC1 domain family member 10C, TBC1 domain family, member 10C | |
Rabbit | |
50 kDa | |
0.1 mL | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (95%), Rat (96%), Bovine (99%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.71mg/mL | |
Western Blot 1:500-1:3000 | |
Q8IV04 | |
TBC1D10C | |
Recombinant protein encompassing a sequence within the center region of human Carabin. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
374403 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction