Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Carabin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531802
Description
Recommended positive controls: A431. Predicted reactivity: Mouse (95%), Rat (96%), Rhesus Monkey (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Carabin is an endogenous inhibitor of calcineurin (see MIM 114105) that also inhibits the Ras (see MIM 190020) signaling pathway through its intrinsic Ras GTPase-activating protein activity (Pan et al., 2007 [PubMed 17230191]).
Specifications
Carabin | |
Polyclonal | |
Unconjugated | |
TBC1D10C | |
1810062O14Rik; AI428527; CARABIN; EBP50-PDZ interactor of 64 kD; EPI64C; TBC1 domain family member 10C; TBC1 domain family, member 10c; TBC1D10C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
374403 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.71 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8IV04 | |
TBC1D10C | |
Recombinant protein encompassing a sequence within the center region of human Carabin. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction