Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carazolol 98.0+%, TCI America™
SDP

Catalog No. C25781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25781G 1 g
C2578200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25781G Supplier TCI America Supplier No. C25781G

Chemical Identifiers

CAS 57775-29-8
Molecular Formula C18H22N2O2
Molecular Weight (g/mol) 298.386
MDL Number MFCD00864588
InChI Key BQXQGZPYHWWCEB-UHFFFAOYSA-N
Synonym 1-(9H-Carbazol-4-yloxy)-3-(isopropylamino)-2-propanol
PubChem CID 71739
IUPAC Name 1-(9H-carbazol-4-yloxy)-3-(propan-2-ylamino)propan-2-ol
SMILES CC(C)NCC(COC1=CC=CC2=C1C3=CC=CC=C3N2)O

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 298.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Carazolol
RTECSNumber UA8685000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.