Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbamazepine 10,11-epoxide, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. p-7187961
Click to view available options
Quantity:
5 mg
25 mg

Carbamazepine 10,11-epoxide is a metabolite of the drug carbamazepine.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
CAS 36507-30-9
Physical Form Neat
Linear Formula C15H12N2O2
MDL Number MFCD00467137
Synonym 1 A,10 b-Dihydro-6 H-dibenzo(b,f)oxireno[d]azepine-6-carboxamide
Type Analytical Standard
Quantity 5 mg
Formula Weight 252.27
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.