Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Carbamazepine
GREENER_CHOICESDP

Catalog No. 409850 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-985-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-985-0 Supplier R&D Systems™ Supplier No. 4098/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of neuronal voltage-gated Na+ channels; anticonvulsant

Inhibitor of neuronal voltage-gated Na+ channels. Exhibits anticonvulsant activity. Potentiates GABA-induced Cl- currents in HEK 293 cells expressing the GABAA receptor α1β2γ2 subtype combination. Can induce autophagy by inhibiting inositol synthesis. Also delays disease onset and prolongs survival in a mouse amyotrophic lateral sclerosis (ALS) model, as well as reducing motor neuron loss and altered muscle morphology.

Chemical Identifiers

CAS 298-46-4
Molecular Formula C15H12N2O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 236.27
Purity >99%
Molecular Weight (g/mol) 236.27
Chemical Name or Material Carbamazepine
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.