Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbazole, 96%

Catalog No. ac108260050
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108260050 5 g Glass Bottle
AC108260250 25 g Plastic bottle
AC108262500 250 g Plastic bottle
AC108260010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC108260050 Supplier Thermo Scientific Chemicals Supplier No. 108260050
Only null left
Add to Cart

CAS: 86-74-8 | C12H9N | 167.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-74-8
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.21
MDL Number MFCD00004960
InChI Key UJOBWOGCFQCDNV-UHFFFAOYSA-N
Synonym carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn
PubChem CID 6854
ChEBI CHEBI:27543
IUPAC Name 9H-carbazole
SMILES N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 240.0°C to 246.0°C
Color Beige-Brown or Beige-Yellow
Boiling Point 355.0°C
Flash Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Quantity 5 g
Beilstein 20,433
Merck Index 15,1791
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in acetic acid- soluble in h2so4,1g/3mL quinoline- 1g/6mL pyridine- 1g/35mL ether,1g/9mL acetone- 1g/120mL benzene,1g/135mL abs. alcohol- slightly soluble in,petroleum ether and
Formula Weight 167.21
Percent Purity 96%
Physical Form Crystalline Powder, Flakes, or Chunks
Chemical Name or Material Carbazole, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
EINECSNumber 201-696-0
RTECSNumber FE3150000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.