Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbetapentane citrate

Catalog No. AAJ6444603
Click to view available options
Quantity:
50 mg
1 g

Sigma receptor agonist | CAS: 23142-01-0 | C26H39NO10 | 525.595 g/mol

Sigma receptor agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23142-01-0
Molecular Formula C26H39NO10
Molecular Weight (g/mol) 525.595
MDL Number MFCD00055697
InChI Key AKJDEXBCRLOVTH-UHFFFAOYSA-N
Synonym carbetapentane citrate, pentoxyverine citrate, loucarbate, pentoxiverine citrate, carbetapentane citrate salt, toclase, toclase citrate, carbetapentone citrate, unii-4sh0mfj5hj
PubChem CID 90010
IUPAC Name 2-[2-(diethylamino)ethoxy]ethyl 1-phenylcyclopentane-1-carboxylate;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCN(CC)CCOCCOC(=O)C1(CCCC1)C2=CC=CC=C2.C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Color White
Quantity 1 g
Merck Index 14,1794
Formula Weight 633.7
Physical Form Powder
Chemical Name or Material Carbetapentane citrate
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 245-449-5
RTECSNumber GY3500000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.