Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carbofuran Monoclonal Antibody (A4H7)
GREENER_CHOICE

Catalog No. PIMA542542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542542 Supplier Invitrogen™ Supplier No. MA542542
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Product has been tested in direct ELISA.

Carbofuran is one of the most toxic carbamate pesticides. It is a systemic insecticide, which means that the plant absorbs it through the roots, and from here the plant distributes it throughout its organs where insecticidal concentrations are attained. Carbofuran exhibits toxicity mediated by the same mechanism as that of the notorious V-series nerve agents and presents a risk to human health.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbofuran
Applications ELISA
Classification Monoclonal
Clone A4H7
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.4
Gene Alias Carbofuran; Furadan
Host Species Mouse
Immunogen Carbofuran coupled with KLH.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.