Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbohydrate Sulfotransferase 15/CHST15 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $610.00
Specifications
Antigen | Carbohydrate Sulfotransferase 15/CHST15 |
---|---|
Concentration | 0.05mg/mL |
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
Carbohydrate Sulfotransferase 15/CHST15 Polyclonal specifically detects Carbohydrate Sulfotransferase 15/CHST15 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Carbohydrate Sulfotransferase 15/CHST15 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Rabbit | |
Human | |
B cell RAG associated protein (GALNAC4S-6ST), B-cell RAG-associated gene protein, BRAGKIAA0598RP11-47G11.1, carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15, carbohydrate sulfotransferase 15, DKFZp781H1369, EC 2.8.2.33, GalNAc4S-6ST, GALNAC4S6ST, hBRAG, MGC34346, N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase | |
CHST15 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
0.05mg/mL | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
51363 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GIVRLRDGLRDRYPVEDYLDLFDLAAHQIHQGLQASSAKEQSKMNTIIIGEASASTMWDNNAWTFFYDNSTDGEPPFLTQDFIHAFQPNARLIVMLRDPVER | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title