Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbohydrate Sulfotransferase 4/CHST4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42 - $666.47
Specifications
| Antigen | Carbohydrate Sulfotransferase 4/CHST4 |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Carbohydrate Sulfotransferase 4/CHST4 Polyclonal specifically detects Carbohydrate Sulfotransferase 4/CHST4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Carbohydrate Sulfotransferase 4/CHST4 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 10164 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TSRMYEFVGLEFLPHLQTWVHNITRGKGMGDHAFHTNARDALNVSQAWRWSLPYEKVSRLQKACGDAMNLLGYRHVRSEQEQRNLLLDLLSTWTVPEQIH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.17, EC 2.8.2.21, galactose/N-acetylglucosamine/N-acetylgalactosamine 6-O-sulfotransferase 3, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 3, glcNAc6ST-2, GlcNAc6ST2, Gn6st-2, GST3, GST-3, HEC-GlcNAc6ST, HECGLCNAC6ST, HEC-GLCNAC-6-ST, High endothelial cells N-acetylglucosamine 6-O-sulfotransferase, L-selectin ligand sulfotransferase, LSSTcarbohydrate sulfotransferase 4, N-acetylglucosamine 6-O-sulfotransferase 2 | |
| CHST4 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title