Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbomer 941, NF, Spectrum™ Chemical

Click to view available options
Quantity:
100 g
2.5 kg
50 lb.
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Carbomer 941, NF is an expanded molecule that will increase continually when more alkyne bonds are presented. It is because of this that carbomers are also called carbon-molecules. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 100% |
Quantity | 45 kg |
Grade | NF |
Loss on Drying | 2% |
Packaging | Fiber Drum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction