Learn More
Carbon black, acetylene, 100% compressed
Description
It is used in optical computing, optical memories, optical signal processing and controlling applications respects, C60 and its derivatives can be widely used in photovoltaic applications, magnetic resonance imaging, anti-HIV drugs, anti-cancer drugs, chemotherapy drugs, cosmetic additives, research and other areas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Color | Black |
Assay Percent Range | >99.9% |
Quantity | 2 x 500 g |
Physical Form | Powder |
Chemical Name or Material | Carbon black |
Frequently Asked Questions (FAQs)
The designation of compression is a generic description of density via mechanical compression of Carbon black when manufactured. Carbon black, acetylene is difficult to densify due to its high structure and the production process. As a result, compression is used to press air out of the powder and allow the density to be recorded. Based on the percentage of compression (50% or 100%), the density can be recorded and used to generate a higher-than-normal density:
- Carbon black, acetylene, 100% compressed (Cat. No. 045527.xx) has a density of 135-160 g/L
- Carbon black, acetylene, 50% compressed, 99.9+% (Cat. No. 039724.xx) has a density of 90-121 g/L
Safety and Handling
Hazard Category | H351 |
Precautionary Statement | P201-P202-P281-P308+P313-P501c |
DOTInformation | DOT Class: 4.1, Packaging Group: III |
EINECSNumber | 215-609-9 |
RTECSNumber | FF5800000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.