Carbon disulfide, 99.9+%, Alfa Aesar

Manufacturer: Alfa Aesar 32472AK

Quantity 250mL

 View more versions of this product

Catalog No. AA32472AK

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75-15-0
    Physical Form Liquid
    Assay Percent Range 99.9+%
    Molecular Formula CS2
    Formula Weight 76.14
    Chemical Name or Material Carbon disulfide

    Specifications

    Formula Weight 76.14
    Quantity 250mL
    Physical Form Liquid
    Density 1.2632
    Assay Percent Range 99.9+%
    Molecular Formula CS2
    CAS 75-15-0
    Chemical Name or Material Carbon disulfide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit