Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon disulfide, ACS, 99.9+%

Catalog No. AA39785AP
Click to view available options
Quantity:
500 mL
2 L
4 x 500 mL

For Use in Electronic Vacuum Tubes, in the Manufacturing of Carbon Tetrachloride, Rayon | CAS: 75-15-0 | CS2 | 76.131 g/mol

It is used in electronic vacuum tubes, in the manufacturing of carbon tetrachloride, rayon, soil disinfectants, as a solvent, manufacture of rayon, cellophane, and carbon tetrachloride, to produce rubber and chemicals. It is used in the manufacture of xanthates. It is also used as a reaction agent for the production of rubber vulcanization. accelerators and soil sterilizers. It is an effective solvent for oils, waxes, sulfur and sulfur compounds. Carbon disulfide and carbon dioxide acts as resources for functional polymers. It is also useful for the synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Color (APHA) 10, Evaporation Residue 0.002%, Hydrogen sulfide (H2S) P.T. (limit about 1.5ppm, Sulfur dioxide (SO2) P.T. (limit about 2.5ppm), H2O 0.05%
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Melting Point -112°C
Color Colorless
Density 1.2632
Boiling Point 46°C
Quantity 500 mL
Flash Point -30°C (-22°F)
Physical Form Liquid
Refractive Index 1.627
Assay Percent Range 99.9+%
Solubility Information Soluble in alcohol,ether,benzene,oil,chloroform and carbon tetrachloride.
Formula Weight 76.14
Grade ACS Reagent
Chemical Name or Material Carbon disulfide, For analysis APS >5 micron
Show More Show Less
Hazard Category H225-H315-H319-H332-H336-H361fd-H372
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P332+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: I
EINECSNumber 200-843-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.