Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon Disulfide, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02002802 Shop All Avantor Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Aluminum Bottle

Catalog No. 02-002-802

Supplier: Avantor 917222

Only null left
Add to Cart
Add to Cart

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Quantity 1 L
Flash Point −30.00°C
Packaging Aluminum Bottle
Specific Gravity 1L = 1.26kg
Formula Weight 76.13
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Carbon Disulfide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.