Alfa Aesar™ Carbon disulfide, HPLC Grade, 99.8+%

Manufacturer: Alfa Aesar™ 38993K2

Quantity 1L

 View more versions of this product

Catalog No. AA38993K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75-15-0
    Molecular Formula CS2
    Formula Weight 76.14
    MDL Number MFCD00011321
    UN Number UN1131
    Merck Index 14,1811
    Chemical Name or Material Carbon disulfide
    Grade HPLC Grade

    Specifications

    Formula Weight 76.14
    Assay Percent Range >99.8%
    Molecular Formula CS2
    Health Hazard 2 GHS H Statement
    H225-H372-H361-H302-H315-H319 Highly flammable liquid and vapor.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of damaging fertility or the unborn child.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Refractive Index 1.6270
    Physical Form Liquid
    MDL Number MFCD00011321
    UN Number UN1131
    Health Hazard 1 Danger
    Density 1.2632g/mL
    TSCA TSCA
    CAS 75-15-0
    Merck Index 14,1811
    DOT Information Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: CARBON DISULFIDE
    Odor Unpleasant
    Flash Point −30° (−22°F)
    Boiling Point 46°C
    Melting Point −112°C
    Quantity 1L
    Chemical Name or Material Carbon disulfide
    Assay ≥99.8%
    Grade HPLC Grade

    GHS H Statement
    H225-H372-H361-H302-H315-H319 Highly flammable liquid and vapor.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of damaging fertility or the unborn child.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit