Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon disulfide, HPLC Grade, 99.8+%

Catalog No. AA38993M1
Click to view available options
Quantity:
1 L
4 x 1 L

CAS: 75-15-0 | CS2 | 76.131 g/mol

Carbon disulfide is used in spectrophotometry, high performance liquid chromatography and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Melting Point °C to 112°C
Color Colorless
Appearance Colorless liquid
Density 1.2632 g/cm3 at 20°C
Boiling Point 46°C
Quantity 4 x 1 L
Flash Point -30°C (-22°F)
Ignition Point 95°C (203°F)
Odor Unpleasant
Physical Form Liquid
Refractive Index 1.627
Assay Percent Range >99.8%
Vapor Pressure 400 hPa (300mm Hg) at 20°C
UN Number UN1131
Merck Index 14,1811
Solubility Information Miscible with alcohol,ether,benzene,oil,chloroform and carbon tetrachloride.
Formula Weight 76.14
Percent Purity ≥99.8%
Grade HPLC
Chemical Name or Material Carbon disulfide
Show More Show Less
Hazard Category H225-H315-H319-H332-H336-H361fd-H372
Hazard Statement GHS H Statement
H225-H372-H361-H302-H315-H319
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: CARBON DISULFIDE
EINECSNumber 200-843-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.