Learn More
Honeywell BW™ Carbon Monoxide Calibration Gas
Honeywell BW™ Carbon Monoxide Calibration Gas is designed to test and calibrate single gas sensors.
Supplier:  Honeywell BW™ CG2M10058
 View more versions of this product
Catalog No. 19-806-898
Only null left
Add to Cart
Description
Description
To be used with BW by Honeywell Analytics Gas Monitors.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
58 L
Honeywell Analytics Gas Monitors including Carbon Monoxide Sensors
35.56 cm
7.62 cm
0.75 kg
Carbon Monoxide
Calibration Gas
(1) Cylinder
100 ppm
10.16 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.