Learn More
Sigma Aldrich Carbon nanotube, single-walled
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Specifications
Specifications
| CAS | 308068-56-6 |
| Melting Point | 3652°C to 3697°C |
| Density | 1.7-1.9 g/cm3 (at 25°C) |
| Quantity | 250 mg |
| Synonym | CHASM(TM); CNT; Signis™ CG200; SWCNT; SWNT; Single wall carbon nanotube |
| Percent Purity | ≥90%carbon basis (≥80%as carbon nanotubes) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.