Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon Standard, Organic, 2000 ppm, Reagents

Catalog No. 01335590
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-335-590 1 L Poly Bottle
01-335-589 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-590 Supplier Reagents Holdings Llc Supplier No. CC2056001A
Only null left
Add to Cart
Add to Cart

Carbon Standard, Organic, 2000 ppm, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Carbon Standard, Organic, 2000 ppm
Packaging Poly Bottle
CAS 7732-18-5, 877-24-7
Specific Gravity 1.002
Physical Form Liquid
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Water Analysis
Quantity 1 L
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.