Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Carbon Steel Magnetic Needle
SCI_ED

Catalog No. S27739 Shop All Eisco Products
Change view
Click to view available options
Includes:
Brass Cup Bearing
Jewel Bearing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S27739 Brass Cup Bearing
S27740 Jewel Bearing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27739 Supplier Eisco™ Supplier No. PH0816C
Add to Cart
Add to Cart

Carbon steel magnetic needle is ideal for magnetism experiments in physics classrooms.

  • Single, magnetic needle.
  • Features bearing options for pivoting.
  • Made of carbon steel.
  • Needle Length: 4 in. (100 mm)
  • Great tool for physics classrooms.

Specifications

Grade 7 to Undergraduate
For Use With (Application) Physics Demonstrations and Experiments
Includes Brass Cup Bearing
Material Carbon Steel
Quantity 1 Ea.
Class Size 1 Lab Group
Product Type Magnetic Needle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.