Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon Tetrachloride, 99%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
56-23-5
CCl4
153.811
MFCD00000785
VZGDMQKNWNREIO-UHFFFAOYSA-N
carbon tetrachloride,perchloromethane,tetrasol,methane, tetrachloro,vermoestricid,benzinoform,necatorine,tetrafinol,carbona,univerm
5943
CHEBI:27385
tetrachloromethane
C(Cl)(Cl)(Cl)Cl
56-23-5
CCl4
153.811
MFCD00000785
VZGDMQKNWNREIO-UHFFFAOYSA-N
carbon tetrachloride,perchloromethane,tetrasol,methane, tetrachloro,vermoestricid,benzinoform,necatorine,tetrafinol,carbona,univerm
5943
CHEBI:27385
tetrachloromethane
C(Cl)(Cl)(Cl)Cl
Melting Point -23°C
Boiling Point 76°C (760mm Hg)
Decomposition Information Decomposes information: >100°C
Quantity 1L
Residue after Evaporation 0.001% max.
Infrared Spectrum Authentic
Color Undesignated
Water 0.01% max.
Density 1.594g/mL
Refractive Index 1.4590 to 1.4610 (20°C, 589nm)
Assay Percent Range 99%
Linear Formula CCl4
UN Number 1846
Beilstein 01, 64
Fieser 03,43; 04,69; 12,551; 14,72; 16,367
Merck Index 15, 1816
Formula Weight 153.82
Physical Form Liquid
Percent Purity ≥98.5%
Grade Extra Pure
Chemical Name or Material Carbon Tetrachloride
Show More Show Less

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Toxic in contact with skin.
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Toxic if swallowed.
Harms public health and the environment by de

Emergency Overview
Toxic by inhalation, in contact with skin and if swallowed. Toxic: danger of serious damage to health by prolonged exposure through inhalation. Suspected human carcinogen. May cause eye, skin, and respiratory tract irritation . May cause central nervous system effects. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Harms public health and the environment by destroying ozone in the upper atmosphere.
NFPA
Health:3 Flammability:0 Instability:0

Signal Word: DANGER!