Learn More
Carbonate, 0.5 M Buffer Solution, pH 9.5, PROMO, Thermo Scientific Chemicals
Description
Applications
Sodium bicarbonate buffer system is important for several different cellular processes. It is one of the major buffering systems used to maintain the pH of mammalian blood and is also important in the acid-base homeostasis of living things, including humans. It tends to maintain a relatively constant plasma pH and counteract any force that would alter it. It is used to neutralize the pH of the chyme leaving the stomach and entering the small intestine.
Solubility
Soluble in water.
Notes
Store at 4°C in tightly closed container. Avoid inhalation. Wash thoroughly after handling. Avoid prolonged or repeated exposure.

Specifications
Specifications
Concentration | 0.5 M |
pH Range | 9.5 |
Quantity | 250 mL |
Type | Carbonate Buffer Solution |
RUO - Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.