Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Carbonate, 0.5M buffer soln., pH 9.5

Catalog No. p-7042754
Click to view available options
Quantity:
250 mL
500 mL

Sodium bicarbonate buffer system is important for several different cellular processes. It is one of the major buffering systems used to maintain the pH of mammalian blood and is also important in the acid-base homeostasis of living things, including humans. It tends to maintain a relatively constant plasma pH and counteract any force that would alter it. It is used to neutralize the pH of the chyme leaving the stomach and entering the small intestine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bicarbonate buffer system is important for several different cellular processes. It is one of the major buffering systems used to maintain the pH of mammalian blood and is also important in the acid-base homeostasis of living things, including humans. It tends to maintain a relatively constant plasma pH and counteract any force that would alter it. It is used to neutralize the pH of the chyme leaving the stomach and entering the small intestine.

Solubility
Soluble in water.

Notes
Store at 4°C in tightly closed container. Avoid inhalation. Wash thoroughly after handling. Avoid prolonged or repeated exposure.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 M buffer soln.
pH Range 9.5
Quantity 250 mL
Type Carbonate buffer soln.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.