Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Carbonic Acid Solution, pH 5.7
SCI_ED

Catalog No. s25234 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Solution
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25234 500 mL Solution
S25234A 4 L Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25234

Carbonic Acid Solution, pH 5.7, CO2, CAS Number-124-38-9, 7732-18-5, carbonica, carbonic acid anhydride, after-damp, carbon oxide, di, dry ice, carbonic anhydride, carbonic acid gas, kohlensaure, khladon 744, kohlendioxyd, 500mL, CHEBI:16526, Gray, Colorless, 1g/mL, CURLTUGMZLYLDI-UHFFFAOYSA-N

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 124-38-9 , 7732-18-5
Molecular Formula CO2
Molecular Weight (g/mol) 44.009
InChI Key CURLTUGMZLYLDI-UHFFFAOYSA-N
Synonym carbonic anhydride, dry ice, carbonic acid gas, carbonic acid anhydride, carbonica, carbon oxide, di, kohlendioxyd, kohlensaure, khladon 744, after-damp
PubChem CID 280
ChEBI CHEBI:16526
IUPAC Name carbon dioxide
SMILES C(=O)=O

Specifications

Color Colorless
Quantity 500 mL
Linear Formula H2CO3
Packaging Solution
Density 1g/mL
Chemical Name or Material Carbonic Acid Solution, pH 5.7
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.