Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase I/CA1 Antibody, Biotin, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP1777400.1MG
Description
Carbonic Anhydrase I/CA1 Polyclonal specifically detects Carbonic Anhydrase I/CA1 in Human samples. It is validated for Western Blot, ELISA.Specifications
Carbonic Anhydrase I/CA1 | |
Polyclonal | |
Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000 | |
CA-I, Car1, Carbonate dehydratase I, carbonic anhydrase 1, Carbonic anhydrase B, carbonic anhydrase ICAB, carbonic dehydratase, EC 4.2.1.1 | |
Goat | |
Delipidation and Defibrination | |
RUO | |
Primary | |
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Goat Serum as well as purified and partially purified Carbonic Anhydrase I [Human Erythrocytes ]. | |
Store at 4C. Do not freeze. | |
IgG |
Western Blot, ELISA | |
Biotin | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
CA1 | |
Carbonic Anhydrase I [Human Erythrocytes] | |
0.1 mg | |
Hypoxia, Lipid and Metabolism | |
759 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction