Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Carbonic Anhydrase I Recombinant Rabbit Monoclonal Antibody (0G10F8)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535508
Description
Immunogen sequence: KVLDALQAIK TKGKRAPFTN FDPSTLLPSS LDFWTYPGSL THPPLYESVT WIICKESISV SSEQLAQFRS LLSNVEGDNA VPMQHNNRPT QPLKGRTVRA SF.
Carbonic anhydrases (CAs) are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. CA1 is closely linked to CA2 and CA3 genes on chromosome 8, and it encodes a cytosolic protein which is found at the highest level in erythrocytes. Variants of this gene have been described in some populations. Multiple alternatively spliced variants, encoding the same protein, have been identified. Transcript variants of CA1 utilizing alternative polyA_sites have been described in literature.
Specifications
Carbonic Anhydrase I | |
Recombinant Monoclonal | |
0.75 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B0BNN3, P13634 | |
Car1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 160-261 of human Carbonic Anhydrase 1 (CA1) (NP_0017291). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
0G10F8 | |
Unconjugated | |
Car1 | |
AW555628; CA I; CA1; ca1 {ECO:0000250; CAB; CA-I; CA-I {ECO:0000250; Car1; Car-1; Carbonate dehydratase I; carbonate dehydratase I {ECO:0000250; carbonic anhydrase 1; carbonic anhydrase 1 {ECO:0000250; Carbonic anhydrase B; carbonic anhydrase I; carbonic anhydrase I {ECO:0000250; epididymis secretory protein Li 11; HEL-S-11; UniProtKB:P00915} | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12346, 310218 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction