Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carbonic Anhydrase I Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519177
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519177 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519177 Supplier Invitrogen™ Supplier No. PA519177
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

Carbonic anhydrases (CAs) are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. CA1 is closely linked to CA2 and CA3 genes on chromosome 8, and it encodes a cytosolic protein which is found at the highest level in erythrocytes. Variants of this gene have been described in some populations. Multiple alternatively spliced variants, encoding the same protein, have been identified. Transcript variants of CA1 utilizing alternative polyA_sites have been described in literature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase I
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CA1
Gene Accession No. P00915
Gene Alias AW555628; CA I; CA1; ca1 {ECO:0000250; CAB; CA-I; CA-I {ECO:0000250; Car1; Car-1; Carbonate dehydratase I; carbonate dehydratase I {ECO:0000250; carbonic anhydrase 1; carbonic anhydrase 1 {ECO:0000250; Carbonic anhydrase B; carbonic anhydrase I; carbonic anhydrase I {ECO:0000250; epididymis secretory protein Li 11; HEL-S-11; UniProtKB:P00915}
Gene Symbols CA1
Host Species Goat
Immunogen Synthetic peptide sequence (QAIKTKGKRAP) corresponding to the internal amino acids of CA1 (aa 166-177).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 759
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.