Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase II/CA2 Antibody (002), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290676PCP
Description
Carbonic Anhydrase II/CA2 Monoclonal antibody specifically detects Carbonic Anhydrase II/CA2 in Mouse samples. It is validated for ELISASpecifications
Carbonic Anhydrase II/CA2 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
002 | |
ELISA | |
CAII, CA-IIEC 4.2.1.1, Car2, Carbonate dehydratase II, carbonic anhydrase 2, carbonic anhydrase B, Carbonic anhydrase C, carbonic anhydrase IICAC, carbonic dehydratase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase II/CA2 (Accession#: AAH55291.1; Met 1-Lys 260). | |
0.1 mL | |
Hypoxia, Lipid and Metabolism | |
760 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction