Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase II/CA2 Antibody (003), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290677V
Description
Carbonic Anhydrase II/CA2 Monoclonal specifically detects Carbonic Anhydrase II/CA2 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Carbonic Anhydrase II/CA2 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
3 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CAII, CA-IIEC 4.2.1.1, Car2, Carbonate dehydratase II, carbonic anhydrase 2, carbonic anhydrase B, Carbonic anhydrase C, carbonic anhydrase IICAC, carbonic dehydratase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase II/CA2 (AAH55291.1; Met1-Lys260). | |
0.1 mL | |
Hypoxia, Lipid and Metabolism | |
760 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction