Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carbonic Anhydrase II Monoclonal Antibody (11A2)
GREENER_CHOICE

Catalog No. PIMA534928
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534928 Supplier Invitrogen™ Supplier No. MA534928
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: THP-1, HL-60, rat liver tissue, human colon tissue, human kidney tissue, human stomach cancer tissue, mouse brain tissue.

CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase II
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 11A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene CA2
Gene Accession No. P00918
Gene Alias AI131712; CA II; CA2; CAC; CAII; CA-II; Car2; Car-2; Carbonate dehydratase II; carbonic anhydrase 2; carbonic anhydrase B; carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282; HEL-76; HEL-S-282; Ltw-5; Lvtw-5
Gene Symbols CA2
Host Species Mouse
Immunogen Recombinant protein within Human Carbonic anhydrase 2 aa 63-240.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 760
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.